
Monoclonal Anti-CD58 antibody produced in mouse

Synonym(s):
Anti-CD58 antigen, (lymphocyte function-associated antigen 3), Anti-LFA3
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing