![Mercaptopurine impurity A CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/163/506/04cc5e39-fcd6-46a5-9998-b5acda9437a3/640/04cc5e39-fcd6-46a5-9998-b5acda9437a3.png)
Mercaptopurine impurity A
![Mercaptopurine impurity A CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/163/506/04cc5e39-fcd6-46a5-9998-b5acda9437a3/640/04cc5e39-fcd6-46a5-9998-b5acda9437a3.png)
Synonym(s):
Hypoxanthine, 6-Hydroxypurine
Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Products
Formula Weight
Manufacturer
Melting Point (°C)
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing