Kirenol
Synonym(s):
(1R)-1-[(2S,4aR,4bS,6S,8R,8aS)-6-hydroxy-8-(hydroxymethyl)-2,4b,8-trimethyl-4,4a,5,6,7,8a,9,10-octahydro-3H-phenanthren-2-yl]ethane-1,2-diol
Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing