Skip to Content
Merck
vibrant-m

Isopromethazine hydrochloride

vibrant-m
Synonym(s):
N,N,β-Trimethyl-10H-phenothiazine-10-ethanamine monohydrochloride, N,N-Dimethyl-2-(10H-phenothiazin-10-yl)propan-1-amine hydrochloride
Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing