![Finasteride for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/334/071/0cec42d1-dd13-4726-84d8-9d6083490d8b/640/0cec42d1-dd13-4726-84d8-9d6083490d8b.png)
Finasteride for peak identification
![Finasteride for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/334/071/0cec42d1-dd13-4726-84d8-9d6083490d8b/640/0cec42d1-dd13-4726-84d8-9d6083490d8b.png)
Synonym(s):
Finasteride, N-(2-methyl-2-propyl)-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamide, N-tert-Butyl-3-oxo-4-aza-5α-androst-1-en-17β-carboxamide, MK-906
Empirical Formula (Hill Notation):
C23H36N2O2
CAS Number:
Molecular Weight:
372.54
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing