![Ethyl sulfate sodium salt solution 1 mg/mL in methanol (as ethyl sulfate), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/374/998/33fcc1ac-8be2-44c2-a1a0-0cca2f5e299a/640/33fcc1ac-8be2-44c2-a1a0-0cca2f5e299a.png)
Ethyl sulfate sodium salt solution
![Ethyl sulfate sodium salt solution 1 mg/mL in methanol (as ethyl sulfate), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/374/998/33fcc1ac-8be2-44c2-a1a0-0cca2f5e299a/640/33fcc1ac-8be2-44c2-a1a0-0cca2f5e299a.png)
Synonym(s):
Ethyl sulfate sodium salt
Empirical Formula (Hill Notation):
C2H5O4SNa
CAS Number:
Molecular Weight:
148.11
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing