Skip to Content
Merck
(−)-Epigallocatechin-3-O-gallate European Pharmacopoeia (EP) Reference Standard

(−)-Epigallocatechin-3-O-gallate

(−)-Epigallocatechin-3-O-gallate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(−)-Epigallocatechin gallate, (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG
Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade