ECO TWEEN® 60
Synonym(s):
TWEEN® 60, Polyethylene glycol sorbitan monostearate, Polyoxyethylene sorbitan monostearate
CAS Number:
Products
Product Category
Brand
Sustainability
Feature
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing