Skip to Content
Merck
dCTP 99% (dNTP), <0.9% (dNDP), solution, 100 mM

dCTP

dCTP 99% (dNTP), <0.9% (dNDP), solution, 100 mM
Synonym(s):
2′-Deoxycytidine 5′-triphosphate disodium salt, dCTP-Na2
Empirical Formula (Hill Notation):
C9H14N3Na2O13P3
CAS Number:
Molecular Weight:
511.12

Products

Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing