
Carbocisteine

Synonym(s):
S-Carboxymethyl-L-cysteine, 3-carboxymethylthio-L-alanine
Empirical Formula (Hill Notation):
C5H9NO4S
CAS Number:
Molecular Weight:
179.19
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing