Skip to Content
Merck
Butylamine analytical standard

Butylamine

Butylamine analytical standard
Synonym(s):
1-Aminobutane, n-Butylamine, Butylamine
Linear Formula:
CH3(CH2)3NH2
CAS Number:
Molecular Weight:
73.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing