Anti-MAP4K2 antibody produced in rabbit
Synonym(s):
Anti-B lymphocyte serine/threonine-protein kinase antibody produced in rabbit, Anti-GC kinase antibody produced in rabbit, Anti-Germinal center kinase antibody produced in rabbit, Anti-MAPK/ERK kinase kinase kinase 2 antibody produced in rabbit, Anti-MEK kinase kinase 2 antibody produced in rabbit, Anti-MEKKK 2 antibody produced in rabbit, Anti-Mitogen-activated protein kinase kinase kinase kinase 2 antibody produced in rabbit, Anti-Rab8-interacting protein antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1