![4-Benzyloxybenzyl alcohol, polymer-bound 100-200 mesh, extent of labeling: 1.5-2.0 mmol/g OH loading, 1 % cross-linked with divinylbenzene](/deepweb/assets/sigmaaldrich/product/structures/262/961/b7cacbcc-d741-47e1-923d-140f76bb73b4/640/b7cacbcc-d741-47e1-923d-140f76bb73b4.png)
4-Benzyloxybenzyl alcohol, polymer-bound
![4-Benzyloxybenzyl alcohol, polymer-bound 100-200 mesh, extent of labeling: 1.5-2.0 mmol/g OH loading, 1 % cross-linked with divinylbenzene](/deepweb/assets/sigmaaldrich/product/structures/262/961/b7cacbcc-d741-47e1-923d-140f76bb73b4/640/b7cacbcc-d741-47e1-923d-140f76bb73b4.png)
Synonym(s):
p-Alkoxybenzyl alcohol, polymer-bound, Wang resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene
Products
Brand
Quality Segment
Functional Group
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing