1-Methylethyl 4-hydroxy-3,5-bis(1-methylethyl)benzoate
Synonym(s):
Isopropyl 4-hydroxy-3,5-diisopropylbenzoate
Empirical Formula (Hill Notation):
C16H24O3
CAS Number:
Molecular Weight:
264.36
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1