![1,1′-Oxybis[3-(2-methoxyphenoxy)-2-propanol] pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/490/164/4f826852-d3cb-4a14-854d-733b4550d51b/640/4f826852-d3cb-4a14-854d-733b4550d51b.png)
1,1′-Oxybis[3-(2-methoxyphenoxy)-2-propanol]
![1,1′-Oxybis[3-(2-methoxyphenoxy)-2-propanol] pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/490/164/4f826852-d3cb-4a14-854d-733b4550d51b/640/4f826852-d3cb-4a14-854d-733b4550d51b.png)
Empirical Formula (Hill Notation):
C20H26O7
Molecular Weight:
378.42
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing