Product Name
Fluorescein isothiocyanate–dextran average mol wt 150,000, (FITC:Glucose = 1:160)
Product Number
46946
Product Brand
sigma
CAS Number
60842-46-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Yellow to Dark Yellow
APPEARANCE (FORM)Powder or Crystals
SOLUBILITY (COLOR)Yellow to Very Dark Yellow
SOLUBILITY (TURBIDITY)Clear to Very Hazy
SOLUBILITY (METHOD)25 MG/ML WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM513 - 523 nm
REMARKS ON LUMINESCENCEEXC. 492 NM, 0.1M PHOSPHATE PH 8.0
Degree of SubstitutionDEGREE OF SUBSTITUTION in FITC/MOL OF GLUCOSE
RemarkAVERAGE MOLECULAR WEIGHT APPROX.150000