Skip to Content
Merck

Skip To

RAB1889

Rat CINC-2 ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Please contact Customer Service for Availability
PLN 3,670.00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
rat ... Cxcl3(171551)
Input:
sample type plasma
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
rat
Storage temp.:
−20°C
Shipped in:
wet ice

PLN 3,670.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 17 pg/mL
standard curve range: 16.38-4000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Cxcl3(171551)

General description

This ELISA antibody pair detects rat CINC-2 alpha.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB1895RAB0118RAB1913
species reactivity

rat

species reactivity

rat

species reactivity

rat

species reactivity

rat

Gene Information

rat ... Cxcl3(171551)

Gene Information

rat ... Ccl11(29397)

Gene Information

rat ... Cxcl1(81503)

Gene Information

rat ... APOBEC3H(64668)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 17 pg/mL
standard curve range: 16.38-4000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.64-400 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 300 pg/mL
standard curve range: 234.4-15000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 26 pg/mL
standard curve range: 24.58-6000 pg/mL

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters