Skip to Content
Merck

B0127

Borax Anhydrous

≥99.0%

Synonym(s):

Sodium tetraborate

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
Na2B4O7
CAS Number:
Molecular Weight:
201.22
EC Number:
215-540-4
UNSPSC Code:
12352302
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

UQGFMSUEHSUPRD-UHFFFAOYSA-N

InChI

1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1

SMILES string

[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

assay

≥99.0%

Looking for similar products? Visit Product Comparison Guide

Application

Buffer component for capillary electrophoresis

Legal Information

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

1330-43-4

CAS No.

1330-43-4

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service