Skip to Content
Merck
All Photos(1)

Documents

MABF3107

Sigma-Aldrich

Anti-ULBP2 Antibody, clone BUMO1

Synonym(s):

ALCAN-alpha, N2DL-2, NKG2D ligand 2, NKG2DL2, Retinoic acid early transcript 1H, UL16-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

BUMO1, monoclonal

mol wt

calculated mol wt 27.37 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
flow cytometry: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... ULBP2(80328)

Specificity

Clone BUMO1 is a mouse monoclonal antibody that detects UL16-binding protein 2 (ULBP2).

Immunogen

p815 murine mastocytoma cells expressing human ULBP2.

Application

Quality Control Testing

Evaluated by Flow Cytometry in C1R cells expressing ULBP2.

Flow Cytometry Analysis: 1.0 µg of this antibody detected ULBP2 in one million C1R cells expressing ULBP2.

Tested Applications

ELISA Analysis: A representative lot detected ULBP2 in ELISA applications (Waldhauer, I., et al. (2006). Cancer Res. 66(5):2520-6; Weil, S., et al. (2017). Front Immunol. 10:8:387).

Flow Cytometry Analysis: A representative lot detected ULBP2 in Flow Cytometry applications (Welte, S.A., et al. (2003). Eur J Immunol. 33(1):194-203; Waldhauer, I., et al. (2006). Cancer Res. 66(5):2520-6; Heinemann, A., et al. (2012). Cancer Res. 72(2):460-71).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

UL16-binding protein 2 (UniProt: Q9BZM5; also known as ALCAN-alpha, NKG2D ligand 2, N2DL-2, NKG2DL2, Retinoic acid early transcript 1H) is encoded by the ULBP2 (also known as N2DL2, RAET1H, UNQ463/PRO791) gene (Gene ID: 80328) in human. The UL16-binding proteins (ULBPs) belong to the family of MHC class I-related molecules that are targets of human CMV glycoprotein. ULBP2 is a GPI-anchor glycoprotein that is synthesized with a signal peptide (aa 1-25) and a propeptide (aa 217-246), which are subsequently cleaved off to produce the mature form. ULBP2 serves as a ligand of the activating immunoreceptor NKG2D and mediates natural kill (NK) cell cytotoxicity. It is detected in cell membrane and endoplasmic reticulum. A secreted for has also been described. Its expression is observed in cancer cells and in the fetus, but not in normal tissues. Tumor cells are reported to release the soluble form of ULBP2 by the action of metalloproteases, however, unlike membrane bound form, it does not down-regulate NKG2D on NK cells. Soluble form of ULBP2 is frequently detected in sera of patients with hematopoietic malignancies. NKG2D, frequently expressed on tumor cells, stimulate tumor immunity. Soluble forms of ULBPs are also reported to induce protein tyrosine phosphorylation, activation of Janus kinase 2, MAP kinase, and PI-3 kinase/Akt signaling. Clone BUMO-1 specifically detects ULBP2 and is suitable for multiple immunodetection applications. (Ref.: Waldhauer, I., and Steinle, A. (2006). Cancer Res 66(5); 2520-2526; Welte, SA., et al. (2003). Eur. J. Immunol. 33(1); 194-203; Sutherland, CL., et al. (2002). J. Immunol. 168(2); 671-679).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service