Skip to Content
Merck
All Photos(1)

Documents

14-592-M

Sigma-Aldrich

PTPMEG-2 Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

Quality Level

description

protein target PTPMEG-2

purified by

(Glutathione-agarose)

suitability

suitable for phosphatase test

UniProt accession no.

Gene Information

human ... PTPN9(5780)

General description

FUNCTION: SwissProt: P43378 # Protein-tyrosine phosphatase that could participate in the transfer of hydrophobic ligands or in functions of the Golgi apparatus.
SIZE: 593 amino acids; 68020 Da
SUBCELLULAR LOCATION: Cytoplasm (Probable).
SIMILARITY: SwissProt: P43378 ## Belongs to the protein-tyrosine phosphatase family. Non-receptor class 3 subfamily. & Contains 1 CRAL-TRIO domain. & Contains 1 tyrosine-protein phosphatase domain.

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-592M when ordering the 250 µg size.
Also available in 10 μg size (cat#14-592) and in bulk (cat# 14-592-K).

Quality

Routinely evaluated by dephosphorylation of 100μM DiFMUP

Physical properties

62.5kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service