- Reduced levels of N'-methyl-2-pyridone-5-carboxamide and lysophosphatidylcholine 16:0 in the serum of patients with intrahepatic cholangiocarcinoma, and the correlation with recurrence-free survival.
Reduced levels of N'-methyl-2-pyridone-5-carboxamide and lysophosphatidylcholine 16:0 in the serum of patients with intrahepatic cholangiocarcinoma, and the correlation with recurrence-free survival.
Oncotarget (2018-01-20)
Kyung-Hee Kim, Jungnam Joo, Boram Park, Sang-Jae Park, Woo Jin Lee, Sung-Sik Han, Tae Hyun Kim, Eun Kyung Hong, Sang Myung Woo, Byong Chul Yoo
PMID29348849
ABSTRACT
We searched for metabolic biomarkers that may predict the prognosis of patients with intrahepatic cholangiocarcinoma (IHCC). To this end, a total of 237 serum samples were obtained from IHCC patients (