Merck
  • Charge-Transfer Analysis of 2p3d Resonant Inelastic X-ray Scattering of Cobalt Sulfide and Halides.
Charge-Transfer Analysis of 2p3d Resonant Inelastic X-ray Scattering of Cobalt Sulfide and Halides.
The journal of physical chemistry. C, Nanomaterials and interfaces (2017-11-25)
Ru-Pan Wang, Boyang Liu, Robert J Green, Mario Ulises Delgado-Jaime, Mahnaz Ghiasi, Thorsten Schmitt, Matti M van Schooneveld, Frank M F de Groot
ABSTRACT
We show that with 2p3d resonant inelastic X-ray scattering (RIXS) we can accurately determine the charge-transfer parameters of CoF
Sigma-Aldrich
Cobalt(II) bromide, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Cobalt sulfide, 99.98% trace metals basis