Merck
  • Irradiation by blue light in the presence of a photoacid confers changes to colony morphology of the plant pathogen Colletotrichum gloeosporioides.
Irradiation by blue light in the presence of a photoacid confers changes to colony morphology of the plant pathogen Colletotrichum gloeosporioides.
Journal of photochemistry and photobiology. B, Biology (2017-07-25)
Ron Simkovitch, Oren Gajst, Einat Zelinger, Oded Yarden, Dan Huppert
ABSTRACT
We used the photoacid 8-hydroxy-1,3,6-pyrenetrisulfonate (HPTS) that converts blue photons to acidic protons in water, with an efficiency of close to 100%, and determined that this treatment conferred changes to colony morphology of the plant pathogen Colletotrichum gloeosporioides. The time elapsed until hyphal collapse is noticed depends on both the laser intensity in mW/cm
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%