Merck
  • Chelating ruthenium phenolate complexes: synthesis, general catalytic activity, and applications in olefin metathesis polymerization.
Chelating ruthenium phenolate complexes: synthesis, general catalytic activity, and applications in olefin metathesis polymerization.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-09-11)
Anna Kozłowska, Maciej Dranka, Janusz Zachara, Eva Pump, Christian Slugovc, Krzysztof Skowerski, Karol Grela
ABSTRACT
Cyclic Ru-phenolates were synthesized, and these compounds were used as olefin metathesis catalysts. Investigation of their catalytic activity pointed out that, after activation with chemical agents, these catalysts promote ring-closing metathesis (RCM), enyne and cross-metathesis (CM) reactions, including butenolysis, with good results. Importantly, these latent catalysts are soluble in neat dicyclopentadiene (DCPD) and show good applicability in ring-opening metathesis polymeriyation (ROMP) of this monomer.
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer