Merck
A cyclic hexapeptide is a potent antagonist of alpha 4 integrins.
Journal of immunology (Baltimore, Md. : 1950) (1997-02-15)
P Vanderslice, K Ren, J K Revelle, D C Kim, D Scott, R J Bjercke, E T Yeh, P J Beck, T P Kogan
ABSTRACT
The alpha 4 integrins mediate leukocyte adhesion to specific counter-receptors, including vascular cell adhesion molecule-1 (VCAM-1), the fibronectin splice variant containing connecting segment 1 (CS1), and mucosal addressin cell adhesion molecule-1. A series of cyclized peptides based on the LDV sequence of CS1 were synthesized and assayed for inhibition of alpha 4 integrin binding. The most potent peptide, C*WLDVC* (where * indicates disulfide-linked residues), inhibited alpha 4 beta 1-dependent binding of lymphocytes to VCAM-1 and CS1 with half-maximal inhibition achieved at 1 to 3 microM of peptide. The peptide proved more potent when the lymphocytes were activated with 1 mM MnCl2; half-maximal inhibition was reached at 0.4 and 0.05 microM for VCAM-1 and CS1, respectively. This represents a 100- to 800-fold increase in potency over a linear CS1 peptide in these same assays. C*WLDVC* also inhibited alpha 4 beta 7-dependent lymphocyte binding to the ligands mucosal addressin cell adhesion molecule-1, VCAM-1 and CS1. Immunoprecipitation of radiolabeled integrin indicated that the peptide could bind alpha 4 beta 1 and alpha 4 beta 7 directly and elute alpha 4 beta 1 from a CS1-conjugated agarose resin. The peptide showed selectivity for alpha 4 integrins in that it effectively inhibited alpha 4 beta 1-dependent, but not alpha 5 beta 1-dependent, binding of cells to intact fibronectin. Due to its small size and potency, C*WLDVC* may serve as a useful tool for the study of alpha 4 integrin biology and the development of small molecule therapeutics.
Sigma-Aldrich
Fibronectin Type III Connecting Segment Fragment 1-25, ≥90% (HPLC)