Merck
  • The wide binding properties of a wheat nonspecific lipid transfer protein. Solution structure of a complex with prostaglandin B2.
The wide binding properties of a wheat nonspecific lipid transfer protein. Solution structure of a complex with prostaglandin B2.
European journal of biochemistry (2000-02-15)
S Tassin-Moindrot, A Caille, J P Douliez, D Marion, F Vovelle
ABSTRACT
The 3D solution structure of wheat nonspecific lipid transfer protein (ns-LTP) complexed with prostaglandin B2, a lipid with both vinyl and hydroxylated groups, has been determined by 1H 2D NMR. The global fold of the protein is close to the previously published structures of wheat, maize, barley and rice ns-LTPs. The ligand is almost completely embedded in the hydrophobic core of the protein. Structure comparisons of free and bound wheat ns-LTP reveal that the binding of prostaglandin B2 hardly affects the global fold of the protein. The structural data on this unusual complex are discussed and compared with other known ns-LTP lipid-complexes.
Sigma-Aldrich
Prostaglandin B2, ≥98%, synthetic
To inquire about this product P5390, please contact your local Merck office or dealer. Contact Technical Service