Merck
  • Fatty acid accumulation in the yeast Sporidiobolus salmonicolor during batch production of gamma-decalactone.
Fatty acid accumulation in the yeast Sporidiobolus salmonicolor during batch production of gamma-decalactone.
FEMS microbiology letters (1997-04-01)
G Feron, L Dufossé, G Mauvais, P Bonnarme, H E Spinnler
ABSTRACT
This paper provides new information about the metabolism of various fatty acids and gamma-decalactone production by yeast. An analysis of the fatty acid composition of the yeast Sporidiobolus salmonicolor during batch production of lactone with ricinoleic acid methyl ester as a precursor showed an accumulation of the gamma-decalactone precursor inside the cells. Electron microscopy of the yeasts showed the presence of large internal inclusions leading to membrane and organelle lysis and, consequently, death of the yeast. S. salmonicolor cultivated with methyl oleate did not produce gamma-decalactone and is viable during the whole culture. Analysis of the long chain fatty acid fraction showed incorporation of methyl oleate.
Sigma-Aldrich
Methyl ricinoleate, ≥99% (GC)
Supelco
Methyl ricinoleate, analytical standard