Merck
  • Regulation of renin release via cyclic ADP-ribose-mediated signaling: evidence from mice lacking CD38 gene.
Regulation of renin release via cyclic ADP-ribose-mediated signaling: evidence from mice lacking CD38 gene.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2013-01-25)
Jing Xiong, Min Xia, Fan Yi, Justine M Abais, Ningjun Li, Krishna M Boini, Pin-Lan Li
ABSTRACT
Despite extensive studies, the intracellular regulatory mechanism of renin production and release is still poorly understood. The present study was designed to test whether CD38-ADP-ribosylcyclase signaling pathway contributes to the regulation of renin production and release, and to examine whether CD38 gene knockout (CD38(-/-)) can change this important renal endocrinal function. ADP-ribosylcyclase activity was estimated utilizing HPLC, cADPR levels from western blot, plasma renin activity from RIA kit, urinary sodium and potassium excretion from fame photometry. The expression of CD38 and the activity of ADP-ribosylcyclase to produce cyclic ADP-ribose (cADPR) were nearly abolished in the kidney from CD38(-/-) mice, indicating that CD38 gene is a major enzyme responsible for the generation of cADPR in vivo. Mice lacking CD38 gene showed increased plasma renin activity (PRA) in either conscious or anesthetized status (P<0.05). Low salt intake significantly increased, but high salt intake significantly decreased renin release in both CD38(+/+) and CD38(-/-) mice. In acute experiments, it was demonstrated that plasma renin activity (PRA) significantly increased upon isoprenaline infusion in CD38(-/-) mice compared to CD38(+/+) mice. Accompanied with such increase in PRA, glomerular filtration rate (GFR), renal blood flow (RBF), urine volume (UV) and sodium excretion (UNaV) more significantly decreased in CD38(-/-) than CD38(+/+) mice. Similarly, more increases in PRA but more decreases in GFR, RBF, UV and UNaV were observed in CD38(-/-) than CD38(+/+) mice when they had a low renal perfusion pressure (RPP). CD38-cADPR-mediated signaling may importantly contribute to the maintenance of low PRA and participate in the regulation of renal hemodynamics and excretory function in mice.
Sigma-Aldrich
Cyclic adenosine diphosphate-ribose, ≥90% (HPLC), lyophilized powder
Sigma-Aldrich
NADase from porcine brain, ≥0.007 unit/mg solid