Merck
  • The catalytic competence of cytochrome P450 in the synthesis of serotonin from 5-methoxytryptamine in the brain: an in vitro study.
The catalytic competence of cytochrome P450 in the synthesis of serotonin from 5-methoxytryptamine in the brain: an in vitro study.
Pharmacological research (2012-10-27)
Anna Haduch, Ewa Bromek, Anna Sadakierska-Chudy, Jacek Wójcikowski, Władysława A Daniel
ABSTRACT
Brain serotonin has been implicated in the pathophysiology of a wide spectrum of psychiatric disorders, as well as in the mechanism of action of psychotropic drugs. The aim of present study was to identify rat cytochrome P450 (CYP) isoforms which can catalyze the O-demethylation of 5-methoxytryptamine to serotonin, and to find out whether that alternative pathway of serotonin synthesis may take place in the brain. The study was conducted on cDNA-expressed CYPs (rat CYP1A1/2, 2A1/2, 2B1, 2C6/11/13, 2D1/2/4/18, 2E1, 3A2 and human CYP2D6), on rat brain and liver microsomes and on human liver microsomes (the wild-type CYP2D6 or the allelic variant 2D6*4*4). Of the rat CYP isoforms studied, CYP2D isoforms were the most efficient in catalyzing the O-demethylation of 5-methoxytryptamine to serotonin, but they were less effective than the human isoform CYP2D6. Microsomes from different brain regions were capable of metabolizing 5-methoxytryptamine to serotonin. The reaction was inhibited by the specific CYP2D inhibitors quinine and fluoxetine. Human liver microsomes of the wild-type CYP2D6 metabolized 5-methoxytryptamine to serotonin more effectively than did the defective CYP2D6*4*4 ones. The obtained results indicate that rat brain CYP2D isoforms catalyze the formation of serotonin from 5-methoxytryptamine, and that the deficit or genetic defect of CYP2D may affect serotonin metabolism in the brain. The results are discussed in the context of their possible physiological and pharmacological significance in vivo.
Sigma-Aldrich
5-Methoxytryptamine, 97%