Merck
  • A simple method for measuring long-range 1H-13C coupling constants in organic molecules.
A simple method for measuring long-range 1H-13C coupling constants in organic molecules.
The Journal of organic chemistry (2007-03-10)
Paloma Vidal, Nuria Esturau, Teodor Parella, Juan F Espinosa
ABSTRACT
This study presents a simple method for measuring long-range heteronuclear coupling constants between protons and proton-bearing carbons. The approach involves recording two conventional 1D-TOCSY experiments in which the offset of the selective proton pulse is set on the low- and high-frequency 13C satellites of an isolated proton signal, Hi. Long-range heteronuclear coupling values between the 13Ci bonded to Hi and the protons Hj,k...n that belong to the same spin system were easily determined from the relative displacement of the relayed Hj,k...n signals in the satellite-selective TOCSY spectra. The sense of the displacement indicated the sign of the coupling constants.
Sigma-Aldrich
trans-Stilbene oxide, 98%
Sigma-Aldrich
cis-Stilbene oxide, 97%