Merck
  • Pressurised fluid extraction of bupirimate and ethirimol from aged soils.
Pressurised fluid extraction of bupirimate and ethirimol from aged soils.
Journal of chromatography. A (2001-06-16)
L J Fitzpatrick, J R Dean
ABSTRACT
This paper assesses the effect of pressurised fluid extraction (PFE) on the recovery of bupirimate and its degradation product, ethirimol from a range of soil types. The analytes were extracted under standard conditions (pressure, 2000 p.s.i.; temperature, 100 degrees C; and, three static flush cycles of 5 min static extraction time each) using a variety of individual and combined solvents. It was found that the recovery of bupirimate was dependent upon the organic matter content of soil.
Product Description
Supelco
Bupirimate, PESTANAL®, analytical standard
Supelco
Ethirimol, PESTANAL®, analytical standard