Merck
  • Synthesis of Bio-Based Methylcyclopentadiene from 2,5-Hexanedione: A Sustainable Route to High Energy Density Jet Fuels.
Synthesis of Bio-Based Methylcyclopentadiene from 2,5-Hexanedione: A Sustainable Route to High Energy Density Jet Fuels.
ChemSusChem (2020-10-21)
Josanne-Dee Woodroffe, Benjamin G Harvey
ABSTRACT
The sustainable, bio-based, platform chemical, 2,5-hexanedione [HD (1)], was efficiently converted to methylcyclopentadiene [MCPD (4)] through a three-step process consisting of intramolecular aldol condensation, catalytic chemoselective hydrogenation, and dehydration. Base-catalyzed aldol condensation of 1 resulted in the formation of 3-methyl-2-cyclopenten-1-one [MCO (2)], which was then converted to 3-methyl-2-cyclopenten-1-ol [MCP (3)] by chemoselective reduction with a ternary Ru catalyst system [RuCl2 (PPh3 )3 /NH2 (CH2 )2 NH2 /KOH]. The hydrogenation proceeded with 96 % chemoselectivity. 3 was then dehydrated over AlPO4 /MgSO4 at 70 °C under reduced pressure to yield 4, which can undergo an ambient temperature [4+2]-Diels-Alder cyclization to generate dimethyldicyclopentadiene (DMDCPD), a commodity chemical useful for the preparation of high-performance fuels and polymers. Through this approach, advanced jet fuels and materials can be conveniently produced from sustainable cellulosic feedstocks.
Sigma-Aldrich
Tris(triphenylphosphine)ruthenium(II) dichloride, 97%
Pricing and availability is not currently available.