Merck
  • Total Salvianolic Acid Injection Prevents Ischemia/Reperfusion-Induced Myocardial Injury Via Antioxidant Mechanism Involving Mitochondrial Respiratory Chain Through the Upregulation of Sirtuin1 and Sirtuin3.
Total Salvianolic Acid Injection Prevents Ischemia/Reperfusion-Induced Myocardial Injury Via Antioxidant Mechanism Involving Mitochondrial Respiratory Chain Through the Upregulation of Sirtuin1 and Sirtuin3.
Shock (Augusta, Ga.) (2018-06-05)
Dan-Dan Huang, Xiao-Hong Wei, Hong-Na Mu, Chun-Shui Pan, Quan Li, Bai-He Hu, Xin Chang, Li Yan, Jing-Yu Fan, Yu-Ying Liu, Jian-Yuan Luo, Jing-Yan Han
ABSTRACT
Sirtuin1 (Sirt1) and Sirtuin3 (Sirt3) are known to participate in regulating mitochondrial function. However, whether Total Salvianolic Acid Injection (TSI) protects against myocardial ischemia/reperfusion (I/R) injury through regulating Sirt1, Sirt3, and mitochondrial respiratory chain complexes is unclear. The aim of this study was to explore the effects of TSI on I/R-induced myocardial injury and the underlying mechanism. Male Sprague-Dawley rats were subjected to 30 min occlusion of the left anterior descending coronary artery followed by 90 min reperfusion with or without TSI treatment (8 mg/kg/h). The results demonstrated that TSI attenuated I/R-induced myocardial injury by the reduced infarct size, recovery of myocardial blood flow, and decreased cardiac apoptosis. Moreover, TSI protected heart from oxidative insults, such as elevation of myeloperoxidase, malondialdehyde, hydrogen peroxide, ROS, as well as attenuated I/R-elicited downregulation of Sirt1, Sirt3, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex 10 (NDUFA10), succinate dehydrogenase complex, subunit A, flavoprotein variant (SDHA), and restoring mitochondrial respiratory chain complexes activity. The in vitro study in H9c2 cells using siRNA transfection further confirmed the critical role of Sirt1 and Sirt3 in the effect of TSI on the expression of NDUFA10 and SDHA. These results demonstrated that TSI attenuated I/R-induced myocardial injury via inhibition of oxidative stress, which was related to the activation of NDUFA10 and SDHA through the upregulation of Sirt1 and Sirt3.