Merck
  • Tim-4 Inhibits NLRP3 Inflammasome via the LKB1/AMPKα Pathway in Macrophages.
Tim-4 Inhibits NLRP3 Inflammasome via the LKB1/AMPKα Pathway in Macrophages.
Journal of immunology (Baltimore, Md. : 1950) (2019-07-03)
Wen Liu, Fuxiang Bai, Hongxing Wang, Yan Liang, Xianhong Du, Cui Liu, Dejian Cai, Jiali Peng, Guangming Zhong, Xiaohong Liang, Chunhong Ma, Lifen Gao
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD), characterized by excessive inflammation and lipid deposition, is one of the most common metabolic liver diseases. The expression of NLRP3 inflammasome in macrophages is significantly increased in NAFLD, and its activation aggravates NAFLD greatly. Tim-4, as the phosphatidylserine (PS) receptor, is expressed highly in macrophages, and macrophage Tim-4 inhibits inflammation under various conditions of immune activation. However, the precise role of Tim-4 in NLRP3 inflammasome regulation and NAFLD pathogenesis remains completely unknown. Using NAFLD mice models, we confirmed that the expression of Tim-4 was increased in liver tissues by Western blot, real-time PCR, immunohistochemistry, and immunofluorescence, especially higher expression in liver macrophages, and Tim-4 knockout mice displayed more severe liver inflammation and hepatic steatosis than controls in NAFLD mice model. In vitro, we found that Tim-4 could inhibit NLRP3 inflammasome activation, and the inhibition was dependent on PS binding domain in the IgV domain. Mechanistically, Tim-4 induced the degradation of NLRP3 inflammasome components through activating AMPKα-mediated autophagy. Specifically, Tim-4 promoted AMPKα phosphorylation by interacting with LKB1 and AMPKα. In addition, PS binding motif was responsible for Tim-4-mediated AMPKα and LKB1 interaction. In conclusion, NAFLD microenvironments upregulate Tim-4 expression in macrophages, and elevated Tim-4, in turn, suppresses NLRP3 inflammasome activation by activating LKB1/AMPKα-mediated autophagy, thereby ameliorating the release of IL-1β and IL-18. Collectively, this study unveils the novel function of Tim-4 in suppressing NLRP3 inflammasome, which would shed new lights on intervention of NAFLD or inflammatory liver diseases by targeting Tim-4.
Sigma-Aldrich
Anti-TIMD4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Anti-TIM-4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution