Skip to Content
Merck
VH032-cyclopropane-F ≥95%

VH032-cyclopropane-F

VH032-cyclopropane-F ≥95%
Synonym(s):
(S,R,S)-AHPC-cyclopropane-F
Empirical Formula (Hill Notation):
C26H33FN4O5S
CAS Number:
Molecular Weight:
532.63

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing