![Paroxetine hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/607/975/314fec23-7d23-43b1-b476-1bb9b7d3c21d/640/314fec23-7d23-43b1-b476-1bb9b7d3c21d.png)
Paroxetine hydrochloride
![Paroxetine hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/607/975/314fec23-7d23-43b1-b476-1bb9b7d3c21d/640/314fec23-7d23-43b1-b476-1bb9b7d3c21d.png)
Synonym(s):
Paroxetine hydrochloride hemihydrate, (3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride hemihydrate
Empirical Formula (Hill Notation):
C19H20FNO3 · HCl · .5 H2O
CAS Number:
Molecular Weight:
374.83
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2