
NovaSeptum® GO Preloaded System with Accurate Volume Syringe, 5-port

Synonym(s):
NovaSeptum® GO Accurate Volume Syringes
Products
Product Category
Brand
Product Line
Agency
Quality Segment
Application
Sterility
Sterilization Method
Showing 1-1 of 1
Compare
Product Number
Description
Pricing