
Mercaptopurine impurity B

Synonym(s):
Adenine, 6-Aminopurine, Vitamin B4
Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing