
L-Methionine-(carboxy-13C,methyl-d3)

Linear Formula:
CD3SCH2CH2CH(NH2)13CO2H
Molecular Weight:
153.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2