
Guggulsterone E

Synonym(s):
(E)-Guggulsterone, (17E)-Pregna-4,17(20)-diene-3,16-dione
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing