Skip to Content
Merck
(Ethylthio)trimethylsilane technical grade, 90%

(Ethylthio)trimethylsilane

(Ethylthio)trimethylsilane technical grade, 90%
Synonym(s):
Ethyl trimethylsilyl sulfide, Trimethylsilyl ethyl sulfide
Linear Formula:
C2H5SSi(CH3)3
CAS Number:
Molecular Weight:
134.32

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing