Skip to Content
Merck
β-Lactamase from Enterobacter cloacae Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)

β-Lactamase from Enterobacter cloacae

β-Lactamase from Enterobacter cloacae Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Synonym(s):
β-Lactamase I, β-Lactamase II, Cephalosporinase, Penicillin amido-β-lactam hydrolase, Penicillinase from Enterobacter cloacae
CAS Number:

Products

Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Technique