Skip to Content
Merck
Bile salts suitable for microbiology, (extracted under controlled conditions from purified fresh bile)

Bile salts

Bile salts suitable for microbiology, (extracted under controlled conditions from purified fresh bile)
Synonym(s):
Bile acids sodium salt, Cholic acid-Deoxycholic acid sodium salt mixture

Products

Product Category
Brand
Color
Manufacturer
Physical Form
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Sterility