Skip to Content
Merck
2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde 97%

2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde

2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde 97%
Synonym(s):
2,2-Difluoro-1,3-benzodioxole-4carbaldehyde, 2,2-Difluorobenzo[d][1,3]dioxole, 4-Formyl-2,2-difluoro-1,3-benzodioxole
Empirical Formula (Hill Notation):
C7H4F2O2
CAS Number:
Molecular Weight:
158.10

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing