Skip to Content
Merck
All Photos(1)

Key Documents

349828

Sigma-Aldrich

2,6-Dihydroxynicotinamide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

impurities

<5% ammonium chloride

mp

249 °C (dec.) (lit.)

SMILES string

NC(=O)c1ccc(O)nc1O

InChI

1S/C6H6N2O3/c7-5(10)3-1-2-4(9)8-6(3)11/h1-2H,(H2,7,10)(H2,8,9,11)

InChI key

UYYJYLJAFBVFJW-UHFFFAOYSA-N

General description

2,6-Dihydroxynicotinamide is an nicotinamide derivative. It can be prepared by the reaction of 1,3-dimethyluracil with malonamide in ethanolic sodium ethoxide, followed by the neutralization of reaction mixture with HCl.[1]

Application

2,6-Dihydroxynicotinamide may be used in chemical synthesis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrimidines. 17. Novel pyrimidine to pyridine transformation reaction. One-step synthesis of pyrido [2, 3-d] pyrimidines.
Hirota K, et al.
The Journal of Organic Chemistry, 46(5), 846-851 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service