Merck
True Gel3D™ Hydrogel Troubleshooting Guide
The TrueGel3D™ hydrogel pre-configured kit (TRUE1) is used to prepare a chemically defined hydrogel with stiffness of ~400 Pa (shear modulus) that can be customized to match that of the native cell environment. The TrueGel3D™ hydrogel is formed by crosslinking of RGD (arginylglycylaspartic acid) degradable polymer with CD cell-degradable crosslinker. The cells are encapsulated during crosslinking and can adhere to the polymer through the RGD peptide and grow within the hydrogel. The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala) that enables cells to spread and migrate by secreting matrix metalloproteases (MMP1, MMP3, MMP7, and MMP9).
The hydrogel can be further dissolved by TrueGel3D™ Enzymatic Cell Recovery Solution (TRUEENZ) for post-culture analysis including RT-PCR, Western blots and other molecular biological studies.
IssuesReasonsSolutions
Weak/low gel formationThiol-reactive groups (polymer) and thiol groups (crosslinker) are not near equimolar concentrationBalance the thiol-reactive groups of polymer with thiol groups of crosslinkers
Excess of peptide with thioglycerol results in few thiol-reactive groupsBalance the thiol groups of peptide and crosslinker with thiol-reactive groups of polymer
Low pH caused by acid in peptide sampleOptimize the buffer pH: For a slow gelling system use a buffer with pH 7.2. For a fast gelling system use a buffer with pH 5.5
Weak network formed because of deteriorated reactive groupsUse fresh aliquots 
Fast gelationpH is too highUse buffer with pH 5.5 to slow down the gelation
Too slow gelation processpH is too slowUse buffer with pH 7.2 to accelerate the gelation
Decrease in cell viabilityLack of adhesion factor in polymerAdd appropriate adhesion peptide or proteins
Acidic environmentUse neutral pH during gel setup
Incomplete dissolution of dextran polymer even after 1 hour incubation with TrueGel3DTM enzymatic cell recovery solutionHigh gel strengthIncrease the incubation period for 2 hours and increase concentration of TrueGel3D™ enzymatic cell recovery solution
Reduced TrueGel3D™ enzymatic cell recovery solution activityUse fresh aliquot of enzyme
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch