Merck
HPLC Analysis of Sugars and Sugar Enantiomers on Astec® CYCLOBOND I 2000
HPLC Analysis of Sugars and Sugar Enantiomers on Astec® CYCLOBOND I 2000 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
Sucrose
≥99.5% (GC)
D-(+)-Maltose monohydrate
≥99% (HPLC), BioXtra
D-(−)-Fructose
≥99% (HPLC), BioXtra
Lactose
tested according to Ph. Eur.
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] water; [B] acetonitrile; (15:85, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
ELSD at 35°C and 3.2 bar nitrogen
injection
10 μL
sample
0.5 mg/mL each in water:acetonitrile, 25:75
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null