Merck
HPLC Analysis of Isocyanate Derivatives on Ascentis® ES-Cyano
HPLC Analysis of Isocyanate Derivatives on Ascentis® ES-Cyano application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® ES-Cyano HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Cyano, 15 cm x 4.6 mm I.D., 5 μm particles (577306-U)
mobile phase
[A] 0.1% ammonium acetate in water; [B] 0.1% ammonium acetate in acetonitrile; (70:30, A:B)
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
5 μL
sample
20 μg/mL in 80:12:8, water: acetonitrile: dimethylsulfoxide
Description
Analysis Note
This application demonstrates the suitability of Ascentis Cyano for the efficient separation of isocyanate derivatives.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany