HPLC Analysis of Epoxy resin, Epikote® 1001 on TSKgel® G3000HXL
Materials
analytical column
Product No.
Description
Pricing
TSKgel® G3000HXL HPLC Column
phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 6 μm particle sizeCONDITIONS
column
TSKgel G3000HXL, 30 cm × 7.8 mm I.D., 6 μm particles (816136)
mobile phase
tetrahydrofuran
flow rate
1 mL/min
detector
UV, 254 nm
Description
Legal Information
Epikote is a registered trademark of Hexion, Inc.
TSKgel is a registered trademark of Tosoh Corporation