GC Analysis of Petitgrain Essential Oil on SUPELCOWAX 10
![GC Analysis of Petitgrain Essential Oil on SUPELCOWAX 10 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/141/135/f1797845-81f5-4d00-b966-5799f07f3845/800/f1797845-81f5-4d00-b966-5799f07f3845.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
75 °C (4 min), then 4 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Petitgrain Oil, Paraguay, neat (W523607)
Description
Legal Information
null